Assay Details
Target Gene Details
Entrez Gene ID: | 145957 |
Gene Name: | neuregulin 4 |
Gene Aliases: |
HRG4 |
Location: |
Chr.15:75935190-76062997 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NRG4 | NM_138573.3 | NP_612640.1 | ||
| XM_017021937.1 | XP_016877426.1 | |||
| XM_017021938.1 | XP_016877427.1 | |||
| XM_017021939.1 | XP_016877428.1 | |||
| XM_017021940.1 | XP_016877429.1 | |||
| XM_017021941.1 | XP_016877430.1 | |||
| XM_017021942.1 | XP_016877431.1 | |||
| XM_017021943.1 | XP_016877432.1 | |||
| XM_017021944.1 | XP_016877433.1 | |||
| XM_017021945.1 | XP_016877434.1 | |||
| XM_017021946.1 | XP_016877435.1 | |||
| XM_017021947.1 | XP_016877436.1 | |||
| XM_017021948.1 | XP_016877437.1 | |||
| AK307362.1 | ||||
| AK311204.1 | ||||
| AK311507.1 | ||||
| AM392364.1 | CAL35829.1 | |||
| BC017568.1 | AAH17568.1 | |||
| BX538100.1 | CAD98015.1 | |||
| CR749519.1 | CAH18333.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2749895 | Chr.15:75415580 - 76472296 on Build GRCh38 | Deletion |
|
| nsv569998 | Chr.15:75940264 - 76258597 on Build GRCh38 | Gain |
|
| esv3636933 | Chr.15:75908231 - 76025938 on Build GRCh38 | Gain |
|
| esv2749898 | Chr.15:75764003 - 77933523 on Build GRCh38 | Deletion |
|
| dgv2622n100 | Chr.15:75904333 - 76022201 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map