Assay Details
Target Gene Details
Entrez Gene ID: | 55466 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member A4 |
Gene Aliases: |
MST104, MSTP104, PRO1472 |
Location: |
Chr.15:78264145-78282196 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJA4 | NM_001130182.1 | NP_001123654.1 | ||
| NM_001130183.1 | NP_001123655.1 | |||
| NM_018602.3 | NP_061072.3 | |||
| XM_005254535.3 | XP_005254592.1 | |||
| XM_006720601.1 | XP_006720664.1 | |||
| AF173388.1 | AAQ13629.1 | |||
| AK090898.1 | BAC03540.1 | |||
| AK096616.1 | BAC04828.1 | |||
| AK098079.1 | BAC05229.1 | |||
| BC021720.2 | AAH21720.1 | |||
| BC031044.1 | AAH31044.2 | |||
| BX648710.1 | CAH10558.1 |
More Information
Set Membership: |
|

Genomic Map