Assay Details
Target Gene Details
Entrez Gene ID: | 374654 |
Gene Name: | kinesin family member 7 |
Gene Aliases: |
ACLS, AGBK, HLS2, JBTS12, UNQ340 |
Location: |
Chr.15:89627970-89663086 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIF7 | NM_198525.2 | NP_940927.2 | ||
| XM_011521531.2 | XP_011519833.1 | |||
| AY358384.1 | AAQ88750.1 | |||
| BC104044.1 | AAI04045.1 | |||
| BC112271.1 | AAI12272.1 | |||
| BC112273.1 | AAI12274.1 | |||
| BC143461.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1659 | Chr.15:89610536 - 89655632 on Build GRCh38 | Deletion |
|
| esv3892721 | Chr.15:89530925 - 89673865 on Build GRCh38 | Gain |
|
| nsv1160317 | Chr.15:89614949 - 89683716 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79232057] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map