Assay Details
Target Gene Details
Entrez Gene ID: | 1136 |
Gene Name: | cholinergic receptor nicotinic alpha 3 subunit |
Gene Aliases: |
LNCR2, NACHRA3, PAOD2 |
Location: |
Chr.15:78593052-78621295 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHRNA3 | NM_000743.4 | NP_000734.2 | ||
| NM_001166694.1 | NP_001160166.1 | |||
| NR_046313.1 | ||||
| BC006114.1 | AAH06114.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1628 | Chr.15:78617170 - 78649604 on Build GRCh38 | Insertion |
|
| nsv1045827 | Chr.15:78615314 - 78712300 on Build GRCh38 | Gain |
|
| nsv457208 | Chr.15:78615690 - 78727268 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map