Assay Details
Target Gene Details
Entrez Gene ID: | 56986 |
Gene Name: | DTW domain containing 1 |
Gene Aliases: |
MDS009 |
Location: |
Chr.15:49620980-49654448 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv569392 | Chr.15:49497216 - 49634103 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76050862] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map