Assay Details
Target Gene Details
Entrez Gene ID: | 51285 |
Gene Name: | RAS like family 12 |
Gene Aliases: |
RIS |
Location: |
Chr.15:65045576-65076654 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RASL12 | XM_005254434.4 | XP_005254491.1 | ||
| XM_017022296.1 | 5 | 665 | XP_016877785.1 |
Target Gene Details
Entrez Gene ID: | 123264 |
Gene Name: | solute carrier family 51 beta subunit |
Gene Aliases: |
OSTB, OSTBETA |
Location: |
Chr.15:65045370-65053396 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC51B | NM_178859.3 | NP_849190.2 | ||
| XM_005254159.4 | XP_005254216.1 | |||
| AK129601.1 | ||||
| AY194242.1 | AAP23992.1 | |||
| BC103842.1 | AAI03843.1 | |||
| BC103843.1 | AAI03844.1 | |||
| BC103844.1 | AAI03845.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2763104 | Chr.15:64923655 - 65128399 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map