Assay Details
Target Gene Details
Entrez Gene ID: | 51187 |
Gene Name: | ribosomal L24 domain containing 1 |
Gene Aliases: |
C15orf15, HRP-L30-iso, L30, RLP24, RPL24, RPL24L, TVAS3 |
Location: |
Chr.15:55181314-55197033 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RSL24D1 | NM_016304.2 | NP_057388.1 | ||
| AB462502.1 | ||||
| AF060926.1 | AAG43138.1 | |||
| AF165521.1 | AAF86651.1 | |||
| AF201949.1 | AAF17241.1 | |||
| AF212226.1 | AAK26249.1 | |||
| AK055713.1 | ||||
| AK315431.1 | ||||
| BC008409.1 | AAH08409.1 | |||
| BC008422.1 | AAH08422.1 | |||
| BC008449.1 | AAH08449.1 | |||
| BC008499.1 | AAH08499.1 | |||
| BC009593.1 | AAH09593.1 | |||
| BC009604.1 | AAH09604.1 | |||
| BC012913.1 | AAH12913.1 | |||
| BC014576.1 | AAH14576.1 | |||
| BC016312.1 | AAH16312.1 | |||
| BC016331.1 | AAH16331.1 | |||
| BC016725.1 | AAH16725.1 | |||
| BC016777.1 | AAH16777.1 | |||
| BC026266.1 | AAH26266.1 | |||
| BC026267.1 | AAH26267.1 | |||
| BC028672.1 | AAH28672.1 | |||
| BC035995.1 | AAH35995.1 | |||
| BC093067.1 | AAH93067.1 | |||
| CR457179.1 | CAG33460.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761899 | Chr.15:53592144 - 55643724 on Build GRCh38 | Gain+Loss |
|
| nsv1039471 | Chr.15:54845408 - 55260357 on Build GRCh38 | Gain |
|
| nsv1043789 | Chr.15:55108674 - 55800256 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs146200419] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map