Assay Details
Target Gene Details
Entrez Gene ID: | 10620 |
Gene Name: | AT-rich interaction domain 3B |
Gene Aliases: |
BDP, DRIL2 |
Location: |
Chr.15:74541177-74598131 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARID3B | NM_001307939.1 | NP_001294868.1 | ||
| NM_006465.3 | NP_006456.1 | |||
| AB208830.1 | BAD92067.1 | |||
| AF116846.1 | AAD09133.1 | |||
| AK298716.1 | ||||
| AK302034.1 | ||||
| BC041792.1 | AAH41792.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map