Assay Details
Target Gene Details
Entrez Gene ID: | 440279 |
Gene Name: | unc-13 homolog C |
Gene Aliases: |
- |
Location: |
Chr.15:53837573-54628608 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UNC13C | XM_017022220.1 | XP_016877709.1 | ||
| XM_017022221.1 | XP_016877710.1 | |||
| XM_017022222.1 | XP_016877711.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv569452 | Chr.15:53788613 - 53864783 on Build GRCh38 | Gain |
|
| esv2761899 | Chr.15:53592144 - 55643724 on Build GRCh38 | Gain+Loss |
|
| esv3636512 | Chr.15:53595200 - 53908380 on Build GRCh38 | Gain |
|
| dgv4531n54 | Chr.15:53598165 - 54164557 on Build GRCh38 | Gain |
|
| esv3636520 | Chr.15:53839973 - 53909788 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77977929] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map