Assay Details
Target Gene Details
Entrez Gene ID: | 23102 |
Gene Name: | TBC1 domain family member 2B |
Gene Aliases: |
- |
Location: |
Chr.15:77994985-78077708 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D2B | NM_015079.5 | NP_055894.6 | ||
| NM_144572.1 | NP_653173.1 | |||
| XM_011521387.2 | XP_011519689.1 | |||
| XM_017022022.1 | XP_016877511.1 | |||
| AB028978.1 | BAA83007.1 | |||
| AK000173.1 | BAA90990.1 | |||
| BC033712.1 | AAH33712.1 | |||
| BC140766.1 | ||||
| BC144482.1 | ||||
| BC152465.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3636977 | Chr.15:77957698 - 78017979 on Build GRCh38 | Gain |
|
| nsv952001 | Chr.15:77970359 - 78013358 on Build GRCh38 | Deletion |
|
| esv3569742 | Chr.15:77736842 - 78048032 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map