Assay Details
Target Gene Details
Entrez Gene ID: | 102 |
Gene Name: | ADAM metallopeptidase domain 10 |
Gene Aliases: |
AD10, AD18, CD156c, CDw156, HsT18717, MADM, RAK, kuz |
Location: |
Chr.15:58595204-58749978 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAM10 | NM_001110.3 | NP_001101.1 | ||
| NM_001320570.1 | NP_001307499.1 | |||
| XM_017021861.1 | XP_016877350.1 | |||
| AF009615.1 | AAC51766.1 | |||
| AK300472.1 | ||||
| BC126253.1 | ||||
| Z48579.1 | CAA88463.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1551 | Chr.15:58599440 - 58644381 on Build GRCh38 | Deletion |
|
| nsv569602 | Chr.15:58622127 - 58672238 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map