Assay Details
Target Gene Details
Entrez Gene ID: | 23604 |
Gene Name: | death associated protein kinase 2 |
Gene Aliases: |
DRP-1, DRP1 |
Location: |
Chr.15:63907036-64046470 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DAPK2 | NM_014326.3 | NP_055141.2 | ||
| XM_011521413.2 | XP_011519715.1 | |||
| XM_011521414.1 | XP_011519716.1 | |||
| XM_011521415.2 | XP_011519717.1 | |||
| XM_011521416.2 | 10 | 1146 | XP_011519718.1 | |
| XM_011521417.1 | XP_011519719.1 | |||
| XM_011521421.2 | XP_011519723.1 | |||
| XM_017022043.1 | 8 | 742 | XP_016877532.1 | |
| XM_017022044.1 | XP_016877533.1 | |||
| XM_017022045.1 | XP_016877534.1 | |||
| XM_017022046.1 | XP_016877535.1 | |||
| XM_017022047.1 | 8 | 783 | XP_016877536.1 | |
| XM_017022048.1 | 7 | 732 | XP_016877537.1 | |
| XM_017022049.1 | 8 | 779 | XP_016877538.1 | |
| XM_017022050.1 | 7 | 584 | XP_016877539.1 | |
| XM_017022051.1 | 7 | 730 | XP_016877540.1 | |
| XM_017022052.1 | 4 | 1266 | XP_016877541.1 | |
| AB018001.1 | BAA88063.1 | |||
| AF052941.1 | AAC35001.1 | |||
| BC114506.1 | AAI14507.1 | |||
| BC114951.1 | AAI14952.1 | |||
| GU056176.1 | 10 | 998 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3636720 | Chr.15:63860455 - 64010781 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73450786] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map