Assay Details
Target Gene Details
Entrez Gene ID: | 10391 |
Gene Name: | coronin 2B |
Gene Aliases: |
CLIPINC |
Location: |
Chr.15:68518373-68727806 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CORO2B | NM_001324014.1 | NP_001310943.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs117486863] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map