Assay Details
Target Gene Details
Entrez Gene ID: | 4947 |
Gene Name: | ornithine decarboxylase antizyme 2 |
Gene Aliases: |
AZ2 |
Location: |
Chr.15:64687574-64703281 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OAZ2 | NM_001301302.1 | NP_001288231.1 | ||
| NM_002537.3 | NP_002528.1 | |||
| AF057297.1 | AAD03265.1 | |||
| AF242520.1 | ||||
| AF242521.1 | AAF99601.1 | |||
| BI597670.1 | ||||
| BX648172.1 | 1 | 3248 | ||
| DA210449.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833038 | Chr.15:64522753 - 64724042 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map