Assay Details
Target Gene Details
Entrez Gene ID: | 2108 |
Gene Name: | electron transfer flavoprotein alpha subunit |
Gene Aliases: |
EMA, GA2, MADD |
Location: |
Chr.15:76216228-76311469 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ETFA | NM_000126.3 | NP_000117.1 | ||
| NM_001127716.1 | NP_001121188.1 | |||
| AA557337.1 | ||||
| AK292979.1 | ||||
| AK300044.1 | ||||
| BC015526.1 | AAH15526.1 | |||
| BC095457.1 | AAH95457.1 | |||
| BT009796.1 | AAP88798.1 | |||
| BU176141.1 | ||||
| J04058.1 | AAA52406.1 | |||
| KU178046.1 | ||||
| KU178047.1 | ||||
| KU178048.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2749895 | Chr.15:75415580 - 76472296 on Build GRCh38 | Deletion |
|
| nsv569998 | Chr.15:75940264 - 76258597 on Build GRCh38 | Gain |
|
| nsv833057 | Chr.15:76075507 - 76293527 on Build GRCh38 | Loss |
|
| esv2749898 | Chr.15:75764003 - 77933523 on Build GRCh38 | Deletion |
|
| nsv527608 | Chr.15:76237926 - 76270397 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map