Assay Details
Target Gene Details
Entrez Gene ID: | 56986 |
Gene Name: | DTW domain containing 1 |
Gene Aliases: |
MDS009 |
Location: |
Chr.15:49620980-49654448 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DTWD1 | NM_001144955.1 | NP_001138427.1 | ||
| NM_020234.5 | NP_064619.2 | |||
| XM_011521815.1 | XP_011520117.1 | |||
| XM_017022419.1 | XP_016877908.1 | |||
| XM_017022420.1 | XP_016877909.1 | |||
| XM_017022421.1 | XP_016877910.1 | |||
| XM_017022422.1 | XP_016877911.1 | |||
| XM_017022423.1 | XP_016877912.1 | |||
| XM_017022424.1 | XP_016877913.1 | |||
| XM_017022425.1 | XP_016877914.1 | |||
| XM_017022426.1 | XP_016877915.1 | |||
| XM_017022427.1 | XP_016877916.1 | |||
| XM_017022428.1 | XP_016877917.1 | |||
| XM_017022429.1 | XP_016877918.1 | |||
| XM_017022430.1 | XP_016877919.1 | |||
| XM_017022431.1 | XP_016877920.1 | |||
| AF168717.1 | AAF87319.1 | |||
| AK290184.1 | ||||
| BC032535.2 | AAH32535.1 | |||
| BC093073.1 | AAH93073.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv569392 | Chr.15:49497216 - 49634103 on Build GRCh38 | Loss |
|

Genomic Map