Assay Details
Target Gene Details
Entrez Gene ID: | 55075 |
Gene Name: | uveal autoantigen with coiled-coil domains and ankyrin repeats |
Gene Aliases: |
NUCLING |
Location: |
Chr.15:70654554-70778823 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UACA | NM_001008224.1 | NP_001008225.1 | ||
| NM_018003.2 | NP_060473.2 | |||
| XM_005254529.4 | XP_005254586.1 | |||
| XM_011521752.2 | XP_011520054.2 | |||
| XM_011521753.2 | XP_011520055.2 | |||
| XM_011521754.2 | XP_011520056.1 | |||
| XM_017022394.1 | XP_016877883.1 | |||
| XM_017022395.1 | XP_016877884.1 | |||
| AB046781.2 | BAB13387.2 | |||
| AF322916.1 | AAG49577.1 | |||
| AK057207.1 | 1 | 509 | ||
| AL834464.1 | CAD39123.1 | |||
| BC113407.1 | ||||
| BC113409.1 | ||||
| BC143266.1 | ||||
| BC143267.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1594 | Chr.15:70645257 - 70677701 on Build GRCh38 | Insertion |
|
| nsv833049 | Chr.15:70582449 - 70687237 on Build GRCh38 | Loss |
|
| nsv1035437 | Chr.15:70644099 - 70746740 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map