Assay Details
Target Gene Details
Entrez Gene ID: | 23168 |
Gene Name: | RTF1 homolog, Paf1/RNA polymerase II complex component |
Gene Aliases: |
GTL7, KIAA0252 |
Location: |
Chr.15:41417104-41483563 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RTF1 | NM_015138.4 | NP_055953.3 | ||
| AK292302.1 | ||||
| BC015052.2 | AAH15052.1 | |||
| BF979715.1 | ||||
| D87440.2 | BAA13382.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1039283 | Chr.15:41328301 - 41453282 on Build GRCh38 | Gain |
|
| nsv569232 | Chr.15:41402179 - 41438048 on Build GRCh38 | Loss |
|
| dgv2580n100 | Chr.15:41267962 - 41453282 on Build GRCh38 | Gain |
|
| nsv524476 | Chr.15:41397034 - 41528002 on Build GRCh38 | Loss |
|
| esv3636244 | Chr.15:41428308 - 41434998 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map