Assay Details
Target Gene Details
Entrez Gene ID: | 23604 |
Gene Name: | death associated protein kinase 2 |
Gene Aliases: |
DRP-1, DRP1 |
Location: |
Chr.15:63907036-64046470 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3636720 | Chr.15:63860455 - 64010781 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map