Assay Details
Target Gene Details
Entrez Gene ID: | 80125 |
Gene Name: | coiled-coil domain containing 33 |
Gene Aliases: |
CT61, HP11097 |
Location: |
Chr.15:74202705-74336472 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC33 | XM_017022629.1 | XP_016878118.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833053 | Chr.15:74067815 - 74262799 on Build GRCh38 | Loss |
|
| nsv471253 | Chr.15:74196866 - 74252807 on Build GRCh38 | Loss |
|
| nsv833054 | Chr.15:74165302 - 74332942 on Build GRCh38 | Loss |
|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116223916] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map