Assay Details
Target Gene Details
Entrez Gene ID: | 64799 |
Gene Name: | IQ motif containing H |
Gene Aliases: |
NYDSP5 |
Location: |
Chr.15:67254772-67501804 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Entrez Gene ID: | 64799 |
Gene Name: | IQ motif containing H |
Gene Aliases: |
NYDSP5 |
Location: |
Chr.15:67254772-67501804 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |