Assay Details
Target Gene Details
Entrez Gene ID: | 54982 |
Gene Name: | ceroid-lipofuscinosis, neuronal 6, late infantile, variant |
Gene Aliases: |
CLN4A, HsT18960, nclf |
Location: |
Chr.15:68206992-68229742 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLN6 | NM_017882.2 | NP_060352.1 | ||
| AK000568.1 | BAA91260.1 | |||
| AK027604.1 | BAB55226.1 | |||
| AK124013.1 | ||||
| AK291175.1 | ||||
| AK293155.1 | ||||
| AK293197.1 | ||||
| BC010849.1 | AAH10849.1 | |||
| BC013130.1 | AAH13130.1 | |||
| BM149330.1 | ||||
| CR457244.1 | CAG33525.1 | |||
| KU178671.1 | ||||
| KU178672.1 | ||||
| KU178673.1 | ||||
| KU178674.1 |
More Information
Set Membership: |
|

Genomic Map