Assay Details
Target Gene Details
Entrez Gene ID: | 79664 |
Gene Name: | interactor of little elongation complex ELL subunit 2 |
Gene Aliases: |
BRCC1, NARG2 |
Location: |
Chr.15:60419609-60479160 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ICE2 | NM_001018089.2 | NP_001018099.1 | ||
| NM_024611.5 | NP_078887.2 | |||
| XM_005254661.1 | XP_005254718.1 | |||
| XM_005254662.1 | XP_005254719.1 | |||
| XM_011522009.1 | XP_011520311.1 | |||
| XM_011522010.1 | XP_011520312.1 | |||
| XM_011522011.2 | XP_011520313.1 | |||
| XM_017022569.1 | XP_016878058.1 | |||
| AF502591.1 | AAQ07460.1 | |||
| AK021958.1 | BAB13948.1 | |||
| AK055752.1 | ||||
| AK095389.1 | ||||
| AK127892.1 | ||||
| AL832046.1 | CAH56220.1 | |||
| AL833122.1 | CAH56202.1 | |||
| AY358546.1 | AAQ88910.1 | |||
| BC013684.1 | AAH13684.1 | |||
| BC071685.1 | AAH71685.1 | |||
| BC107427.1 | AAI07428.1 | |||
| BC140894.1 | ||||
| BX647873.1 | CAH56197.1 | |||
| CR749852.1 | CAH18700.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv569609 | Chr.15:60417705 - 60502450 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114307940] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map