Assay Details
Target Gene Details
Entrez Gene ID: | 80381 |
Gene Name: | CD276 molecule |
Gene Aliases: |
4Ig-B7-H3, B7-H3, B7H3, B7RP-2 |
Location: |
Chr.15:73683966-73714518 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD276 | NM_001024736.1 | NP_001019907.1 | ||
| NM_025240.2 | NP_079516.1 | |||
| XM_005254700.4 | XP_005254757.1 | |||
| XM_011522095.2 | XP_011520397.1 | |||
| XM_011522096.2 | XP_011520398.1 | |||
| XM_017022638.1 | XP_016878127.1 | |||
| AJ583696.1 | ||||
| AK074849.1 | BAC11243.1 | |||
| AK075549.1 | BAC11692.1 | |||
| AK291721.1 | ||||
| AK292711.1 | ||||
| AK296358.1 | ||||
| AK298522.1 | ||||
| AK309728.1 | ||||
| AY358343.1 | AAQ88709.1 | |||
| BC062581.1 | AAH62581.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map