Assay Details
Target Gene Details
Entrez Gene ID: | 83660 |
Gene Name: | talin 2 |
Gene Aliases: |
ILWEQ |
Location: |
Chr.15:62390527-62844631 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TLN2 | XM_005254708.4 | XP_005254765.1 | ||
| XM_005254710.4 | XP_005254767.1 | |||
| XM_005254711.4 | XP_005254768.1 | |||
| XM_005254714.3 | XP_005254771.1 | |||
| AB621808.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4567n54 | Chr.15:62413995 - 62428164 on Build GRCh38 | Loss |
|
| nsv833031 | Chr.15:62330187 - 62509069 on Build GRCh38 | Loss |
|
| nsv1035441 | Chr.15:62406037 - 63349997 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs111370369] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map