Assay Details
Target Gene Details
Entrez Gene ID: | 23060 |
Gene Name: | zinc finger protein 609 |
Gene Aliases: |
- |
Location: |
Chr.15:64460350-64686068 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF609 | XM_011521386.2 | XP_011519688.1 | ||
| XM_017022021.1 | XP_016877510.1 | |||
| BC142647.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs72741343] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map