Assay Details
Target Gene Details
Entrez Gene ID: | 390616 |
Gene Name: | ankyrin repeat domain 34C |
Gene Aliases: |
- |
Location: |
Chr.15:79282804-79298239 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD34C | NM_001146341.1 | NP_001139813.1 | ||
| AK127037.1 |

Genomic Map