Assay Details
Target Gene Details
Entrez Gene ID: | 400451 |
Gene Name: | family with sequence similarity 174 member B |
Gene Aliases: |
- |
Location: |
Chr.15:92617447-92734219 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM174B | NM_207446.2 | NP_997329.2 | ||
| AK075564.1 | BAC11703.1 | |||
| BC045729.1 | AAH45729.1 | |||
| BC060873.1 | AAH60873.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1674 | Chr.15:92637781 - 92671702 on Build GRCh38 | Insertion |
|
| nsv1037972 | Chr.15:92261678 - 92727531 on Build GRCh38 | Loss |
|

Genomic Map