Assay Details
Target Gene Details
Entrez Gene ID: | 9133 |
Gene Name: | cyclin B2 |
Gene Aliases: |
HsT17299 |
Location: |
Chr.15:59105085-59125045 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCNB2 | NM_004701.3 | NP_004692.1 | ||
| AB020981.1 | BAA78387.1 | |||
| AF002822.1 | AAD09309.1 | |||
| AF087910.1 | AAP97207.1 | |||
| AK001404.1 | ||||
| AK222611.1 | BAD96331.1 | |||
| AL080146.1 | CAB45739.1 | |||
| AM851054.1 | ||||
| BC105086.1 | AAI05087.1 | |||
| BC105112.1 | AAI05113.1 | |||
| BT019458.1 | AAV38265.1 | |||
| BU934242.1 | ||||
| CR533527.1 | CAG38558.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527307 | Chr.15:58811129 - 59726288 on Build GRCh38 | Gain |
|
| esv3636638 | Chr.15:59109966 - 59112994 on Build GRCh38 | Loss |
|
| nsv833025 | Chr.15:59043949 - 59229977 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map