Assay Details
Target Gene Details
Entrez Gene ID: | 123355 |
Gene Name: | leucine rich repeat containing 28 |
Gene Aliases: |
- |
Location: |
Chr.15:99251362-99390732 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv570716 | Chr.15:99086672 - 99376127 on Build GRCh38 | Gain |
|
| nsv1055008 | Chr.15:98933915 - 99479656 on Build GRCh38 | Gain |
|
| esv3892741 | Chr.15:98979139 - 99618239 on Build GRCh38 | Gain |
|
| nsv1045124 | Chr.15:98961614 - 99307005 on Build GRCh38 | Gain |
|
| esv3637424 | Chr.15:99216374 - 99369158 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74764679] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map