Assay Details
Target Gene Details
Entrez Gene ID: | 25942 |
Gene Name: | SIN3 transcription regulator family member A |
Gene Aliases: |
- |
Location: |
Chr.15:75369379-75455819 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIN3A | NM_001145357.1 | 21 | 5412 | NP_001138829.1 |
| NM_001145358.1 | 21 | 5587 | NP_001138830.1 | |
| NM_015477.2 | 21 | 5489 | NP_056292.1 | |
| XM_006720465.3 | 21 | 5295 | XP_006720528.1 | |
| XM_006720466.3 | 22 | 5685 | XP_006720529.1 | |
| XM_006720467.3 | 23 | 5503 | XP_006720530.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2621n100 | Chr.15:75101872 - 75382048 on Build GRCh38 | Gain |
|
| esv3636926 | Chr.15:75363580 - 75378339 on Build GRCh38 | Gain |
|
| esv2749894 | Chr.15:75326873 - 75749895 on Build GRCh38 | Deletion |
|
| nsv1615 | Chr.15:75351396 - 75377578 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map