Assay Details
Target Gene Details
Entrez Gene ID: | 7090 |
Gene Name: | transducin like enhancer of split 3 |
Gene Aliases: |
ESG, ESG3, GRG3, HsT18976 |
Location: |
Chr.15:70047790-70097935 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
More Information
Set Membership: |
|

Genomic Map