Assay Details
Target Gene Details
Entrez Gene ID: | 283777 |
Gene Name: | family with sequence similarity 169 member B |
Gene Aliases: |
- |
Location: |
Chr.15:98437162-98514382 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM169B | NM_182562.2 | NP_872368.2 | ||
| AK097062.1 | BAC04940.1 |

Genomic Map