Assay Details
Target Gene Details
Entrez Gene ID: | 290 |
Gene Name: | alanyl aminopeptidase, membrane |
Gene Aliases: |
APN, CD13, GP150, LAP1, P150, PEPN |
Location: |
Chr.15:89784895-89814852 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANPEP | NM_001150.2 | NP_001141.2 | ||
| XM_005254892.4 | XP_005254949.1 | |||
| XM_011521473.1 | XP_011519775.1 | |||
| AB209918.1 | BAD93155.1 | |||
| AK298134.1 | ||||
| AK298242.1 | ||||
| AK298339.1 | ||||
| AK300946.1 | ||||
| BC058928.1 | AAH58928.1 | |||
| M22324.1 | AAA51719.1 | |||
| X13276.1 | CAA31640.1 |
More Information
Set Membership: |
|

Genomic Map