Assay Details
Target Gene Details
Entrez Gene ID: | 3419 |
Gene Name: | isocitrate dehydrogenase 3 (NAD(+)) alpha |
Gene Aliases: |
- |
Location: |
Chr.15:78149326-78170542 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IDH3A | NM_005530.2 | NP_005521.1 | ||
| XM_005254336.2 | XP_005254393.1 | |||
| XM_017022118.1 | XP_016877607.1 | |||
| XM_017022119.1 | XP_016877608.1 | |||
| XM_017022120.1 | XP_016877609.1 | |||
| AK093531.1 | ||||
| AK222973.1 | BAD96693.1 | |||
| AK296002.1 | ||||
| AK299970.1 | ||||
| AK315963.1 | ||||
| AK316051.1 | ||||
| AL442090.1 | CAC09449.1 | |||
| BC021967.2 | AAH21967.1 | |||
| CR749548.1 | CAH18348.1 | |||
| U07681.1 | AAA85639.1 |
More Information
Set Membership: |
|

Genomic Map