Assay Details
Target Gene Details
Entrez Gene ID: | 9325 |
Gene Name: | thyroid hormone receptor interactor 4 |
Gene Aliases: |
ASC-1, ASC1, HsT17391, SMABF1, ZC2HC5 |
Location: |
Chr.15:64387804-64455303 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRIP4 | NM_001321924.1 | NP_001308853.1 | ||
| NM_016213.4 | NP_057297.2 | |||
| NR_135855.1 | ||||
| AK314319.1 | ||||
| BC012448.1 | AAH12448.1 | |||
| BG166305.1 | ||||
| L40371.1 | AAC41738.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892709 | Chr.15:64335104 - 64418975 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map