Assay Details
Target Gene Details
Entrez Gene ID: | 55930 |
Gene Name: | myosin VC |
Gene Aliases: |
- |
Location: |
Chr.15:52192318-52295798 on Build GRCh38 |
Assay Gene Location: | Within Intron 35 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYO5C | NM_018728.3 | NP_061198.2 | ||
| XM_011521781.2 | XP_011520083.1 | |||
| XM_017022408.1 | XP_016877897.1 | |||
| XM_017022409.1 | XP_016877898.1 | |||
| XM_017022410.1 | XP_016877899.1 | |||
| AF272390.1 | AAF78783.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471244 | Chr.15:52013266 - 52219268 on Build GRCh38 | Gain |
|
| nsv833005 | Chr.15:52097672 - 52249133 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map