Assay Details
Target Gene Details
Entrez Gene ID: | 4261 |
Gene Name: | class II, major histocompatibility complex, transactivator |
Gene Aliases: |
C2TA, CIITAIV, MHC2TA, NLRA |
Location: |
Chr.16:10866208-10941562 on Build GRCh38 |
Assay Gene Location: | Within Intron 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CIITA | XM_006720880.3 | XP_006720943.2 | ||
| XM_011522485.2 | XP_011520787.1 | |||
| XM_011522487.2 | XP_011520789.1 | |||
| XM_011522488.2 | XP_011520790.1 | |||
| XM_011522489.2 | XP_011520791.1 | |||
| XM_011522490.2 | XP_011520792.1 | |||
| XM_011522492.2 | XP_011520794.1 | |||
| XM_011522493.2 | XP_011520795.1 | |||
| XM_011522494.2 | XP_011520796.1 | |||
| XM_011522495.2 | XP_011520797.1 | |||
| XM_011522496.2 | XP_011520798.1 | |||
| U18259.1 | 22 | 6593 | AAA88861.1 |
Target Gene Details
Entrez Gene ID: | 28955 |
Gene Name: | Dexi homolog |
Gene Aliases: |
MYLE |
Location: |
Chr.16:10928891-10943007 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DEXI | NM_014015.3 | NP_054734.2 | ||
| XM_011522455.2 | XP_011520757.1 | |||
| XM_017023171.1 | XP_016878660.1 | |||
| AF108145.2 | AAC83382.2 | |||
| AF129929.1 | ||||
| BC001083.2 | AAH01083.2 | |||
| BC037569.1 | AAH37569.1 | |||
| BC052989.1 | AAH52989.1 | |||
| BC071683.1 | AAH71683.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv571452 | Chr.16:10907520 - 10936885 on Build GRCh38 | Loss |
|
| nsv1040428 | Chr.16:10145408 - 11362136 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map