Assay Details
Target Gene Details
Entrez Gene ID: | 3163 |
Gene Name: | heme oxygenase 2 |
Gene Aliases: |
HO-2 |
Location: |
Chr.16:4474697-4510347 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HMOX2 | NM_001127204.1 | NP_001120676.1 | ||
| NM_001127206.2 | NP_001120678.1 | |||
| NM_001286267.1 | NP_001273196.1 | |||
| NM_001286268.1 | NP_001273197.1 | |||
| NM_001286269.1 | NP_001273198.1 | |||
| NM_001286271.1 | NP_001273200.1 | |||
| NM_002134.3 | NP_002125.3 | |||
| XM_017023196.1 | XP_016878685.1 | |||
| XM_017023197.1 | XP_016878686.1 | |||
| AK222629.1 | BAD96349.1 | |||
| AK313660.1 | ||||
| BC002396.2 | AAH02396.1 | |||
| BE561721.1 | ||||
| BI225653.1 | ||||
| BM905872.1 | ||||
| BP329113.1 | ||||
| DA141643.1 | ||||
| DA694137.1 | ||||
| DB184091.1 | ||||
| S34389.1 | AAB22110.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160332 | Chr.16:4367643 - 4486554 on Build GRCh38 | Duplication |
|
| nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map