Assay Details
Target Gene Details
Entrez Gene ID: | 85407 |
Gene Name: | naked cuticle homolog 1 |
Gene Aliases: |
Naked1 |
Location: |
Chr.16:50548330-50649249 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKD1 | NM_033119.4 | NP_149110.1 | ||
| XM_011523399.2 | XP_011521701.1 | |||
| XM_017023804.1 | XP_016879293.1 | |||
| AB062886.1 | BAB70500.1 | |||
| AF289584.1 | AAL55768.1 | |||
| AF358135.1 | AAK57484.1 | |||
| AK314930.1 | ||||
| AY061883.1 | AAL32374.1 | |||
| BC051288.1 | AAH51288.1 |
More Information
Set Membership: |
|

Genomic Map