Assay Details
Target Gene Details
Entrez Gene ID: | 9516 |
Gene Name: | lipopolysaccharide induced TNF factor |
Gene Aliases: |
PIG7, SIMPLE, TP53I7 |
Location: |
Chr.16:11547722-11636377 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LITAF | NM_001136472.1 | NP_001129944.1 | ||
| NM_001136473.1 | NP_001129945.1 | |||
| NM_004862.3 | NP_004853.2 | |||
| NR_024320.1 | ||||
| XM_006720982.2 | XP_006721045.1 | |||
| XM_006720984.3 | XP_006721047.1 | |||
| XM_006720985.3 | XP_006721048.1 | |||
| XM_011522754.2 | XP_011521056.1 | |||
| XM_017023896.1 | XP_016879385.1 | |||
| AB034747.1 | BAB32547.1 | |||
| AF010312.1 | AAC39530.1 | |||
| AK095955.1 | ||||
| AK130788.1 | ||||
| AK314519.1 | ||||
| BC000053.2 | ||||
| BC008309.1 | AAH08309.1 | |||
| BC016491.1 | AAH16491.1 | |||
| BC039840.1 | AAH39840.1 | |||
| BC096063.1 | AAH96063.1 | |||
| BC096065.1 | AAH96065.1 | |||
| BC096066.1 | AAH96066.1 | |||
| BC101401.1 | AAI01402.1 | |||
| BC101402.1 | AAI01403.1 | |||
| BC101969.1 | AAI01970.1 | |||
| BI858484.1 | ||||
| BX537543.1 | CAD97778.1 | |||
| DA955452.1 | ||||
| U77396.1 | AAB36550.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524676 | Chr.16:11475598 - 11701723 on Build GRCh38 | Loss |
|
| nsv457409 | Chr.16:11449038 - 11728210 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72779166] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map