Assay Details
Target Gene Details
Entrez Gene ID: | 23406 |
Gene Name: | coactosin like F-actin binding protein 1 |
Gene Aliases: |
CLP |
Location: |
Chr.16:84565596-84618096 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COTL1 | NM_021149.3 | NP_066972.1 | ||
| AK026033.1 | ||||
| AK315675.1 | ||||
| BC010039.2 | AAH10039.1 | |||
| BC010884.1 | AAH10884.1 | |||
| BC016702.1 | AAH16702.1 | |||
| BC042970.1 | AAH42970.1 | |||
| BC053682.1 | AAH53682.1 | |||
| BT006968.1 | AAP35614.1 | |||
| BX395081.2 | ||||
| DA968872.1 | ||||
| L54057.1 | AAA88022.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1055503 | Chr.16:84577279 - 84613367 on Build GRCh38 | Gain |
|
| nsv471109 | Chr.16:84579799 - 84639929 on Build GRCh38 | Loss |
|
| nsv833311 | Chr.16:84489192 - 84663857 on Build GRCh38 | Loss |
|
| nsv1064308 | Chr.16:84461124 - 84592674 on Build GRCh38 | Loss |
|
| dgv550e214 | Chr.16:84569011 - 84602293 on Build GRCh38 | Loss |
|
| dgv3058n100 | Chr.16:84574421 - 84612845 on Build GRCh38 | Loss |
|
| nsv483044 | Chr.16:84166395 - 87066394 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map