Assay Details
Target Gene Details
Entrez Gene ID: | 4261 |
Gene Name: | class II, major histocompatibility complex, transactivator |
Gene Aliases: |
C2TA, CIITAIV, MHC2TA, NLRA |
Location: |
Chr.16:10866208-10941562 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CIITA | XM_006720880.3 | XP_006720943.2 | ||
| XM_011522484.2 | XP_011520786.1 | |||
| XM_011522485.2 | XP_011520787.1 | |||
| XM_011522486.2 | XP_011520788.1 | |||
| XM_011522487.2 | XP_011520789.1 | |||
| XM_011522489.2 | XP_011520791.1 | |||
| XM_011522491.2 | XP_011520793.1 | |||
| XM_011522494.2 | XP_011520796.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3637917 | Chr.16:10867298 - 10904150 on Build GRCh38 | Loss |
|
| nsv1040428 | Chr.16:10145408 - 11362136 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map