Assay Details
Target Gene Details
Entrez Gene ID: | 1071 |
Gene Name: | cholesteryl ester transfer protein |
Gene Aliases: |
BPIFF, HDLCQ10 |
Location: |
Chr.16:56961923-56983844 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CETP | NM_000078.2 | NP_000069.2 | ||
| NM_001286085.1 | NP_001273014.1 | |||
| AA999853.1 | ||||
| AK291586.1 | ||||
| AK297699.1 | ||||
| BC025739.1 | AAH25739.1 | |||
| BT020064.1 | AAV38867.1 | |||
| KU177972.1 | ||||
| KU177973.1 | ||||
| M30185.1 | AAA51977.1 | |||
| M83573.1 | AAB59388.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059098 | Chr.16:56733938 - 57028336 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map