Assay Details
Target Gene Details
Entrez Gene ID: | 10428 |
Gene Name: | craniofacial development protein 1 |
Gene Aliases: |
BCNT, BUCENTAUR, CENP-29, CP27, SWC5, Yeti, p97 |
Location: |
Chr.16:75293710-75433512 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFDP1 | NM_006324.2 | NP_006315.1 | ||
| AB009285.1 | BAA31867.1 | |||
| AK226031.1 | ||||
| AK313948.1 | ||||
| BC000991.2 | AAH00991.1 | |||
| BT009819.1 | AAP88821.1 | |||
| CR542111.1 | CAG46908.1 | |||
| D86549.1 | BAA20070.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833283 | Chr.16:75268277 - 75358410 on Build GRCh38 | Loss |
|
| esv3582321 | Chr.16:75232709 - 75427727 on Build GRCh38 | Loss |
|
| nsv1879 | Chr.16:75310682 - 75345394 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75724671] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map