Assay Details
Target Gene Details
Entrez Gene ID: | 79728 |
Gene Name: | partner and localizer of BRCA2 |
Gene Aliases: |
FANCN, PNCA3 |
Location: |
Chr.16:23603162-23641357 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PALB2 | NM_024675.3 | NP_078951.2 | ||
| XM_011545946.2 | XP_011544248.1 | |||
| XM_011545947.2 | XP_011544249.1 | |||
| XM_011545948.2 | XP_011544250.1 | |||
| XM_017023671.1 | XP_016879160.1 | |||
| XM_017023672.1 | XP_016879161.1 | |||
| XM_017023673.1 | XP_016879162.1 | |||
| AK025469.1 | BAB15140.1 | |||
| AK299152.1 | ||||
| AL834425.1 | CAD39086.1 | |||
| BC044254.2 | AAH44254.1 | |||
| BX647125.1 | ||||
| BX647130.1 | ||||
| CR749637.1 | CAH18431.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs113580717] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map