Assay Details
Target Gene Details
Entrez Gene ID: | 124359 |
Gene Name: | chromodomain protein, Y-like 2 |
Gene Aliases: |
PCCP1 |
Location: |
Chr.16:80597899-80805043 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDYL2 | NM_152342.3 | NP_689555.2 | ||
| XM_011522866.1 | XP_011521168.1 | |||
| XM_011522867.2 | XP_011521169.1 | |||
| AK096185.1 | BAC04720.1 | |||
| AX781670.1 | CAE11965.1 | |||
| AX781671.1 | CAE11953.1 | |||
| AY273798.1 | AAP83842.1 | |||
| BC069440.1 | AAH69440.1 | |||
| BC100803.1 | AAI00804.1 | |||
| BC100804.1 | AAI00805.1 | |||
| BC100805.1 | AAI00806.1 | |||
| BC100806.1 | AAI00807.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv542e214 | Chr.16:80565677 - 80654516 on Build GRCh38 | Gain |
|
| dgv3043n100 | Chr.16:80602287 - 80632882 on Build GRCh38 | Gain |
|
| esv3639277 | Chr.16:80588355 - 80633154 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map