Assay Details
Target Gene Details
Entrez Gene ID: | 1339 |
Gene Name: | cytochrome c oxidase subunit 6A2 |
Gene Aliases: |
COX6AH, COXVIAH |
Location: |
Chr.16:31427731-31428428 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COX6A2 | NM_005205.3 | NP_005196.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833194 | Chr.16:31346858 - 31517224 on Build GRCh38 | Loss |
|
| nsv1066280 | Chr.16:31385143 - 31440746 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map