Assay Details
Target Gene Details
Entrez Gene ID: | 54925 |
Gene Name: | zinc finger and SCAN domain containing 32 |
Gene Aliases: |
HCCS-5, ZNF434 |
Location: |
Chr.16:3382081-3401065 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044147 | Chr.16:3149392 - 3392037 on Build GRCh38 | Loss |
|
| nsv1054969 | Chr.16:3358911 - 3391256 on Build GRCh38 | Loss |
|
| dgv2695n100 | Chr.16:3347901 - 3424504 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map