Assay Details
Target Gene Details
Entrez Gene ID: | 2521 |
Gene Name: | FUS RNA binding protein |
Gene Aliases: |
ALS6, ETM4, FUS1, HNRNPP2, POMP75, TLS |
Location: |
Chr.16:31179444-31194871 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FUS | NM_001170634.1 | 15 | 3704 | NP_001164105.1 |
| NM_001170937.1 | 15 | 3695 | NP_001164408.1 | |
| NM_004960.3 | 15 | 3707 | NP_004951.1 | |
| NR_028388.2 | 14 | 3672 |
More Information
Set Membership: |
|

Genomic Map